Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
cis-1,2-Cyclohexanedimethanol
cis-1,2-Cyclohexanedimethanol 95%
cis-1,2-Cyclohexanedimethanol 95%
cis-1,2-Cyclohexanedimethanol 95%
Supplier:
Apollo Scientific
Certificates
Expand image
cis-1,2-Cyclohexanedimethanol 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
15753-50-1
:
Formula:
C₈H₁₆O₂
MDL Number:
MFCD00062987
:
144.21 g/mol
Storage Temperature:
Ambient