1-Cyclohexyl-2-pyrrolidone 95%
Catalog # APOSOR936238-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:167.25 g/mol
Storage Temperature:Ambient
CAS Number:6837-24-7
Formula:C₁₀H₁₇NO
MDL Number:MFCD00003191