3-Methyl-2-cyclohexen-1-one 98%
Catalog # APOSOR932249-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:110.16 g/mol
Boiling Pt:197 °C
Density:0.97 g/cm³ (25 °C)
Storage Temperature:Ambient
CAS Number:1193-18-6
Formula:C₇H₁₀O
MDL Number:MFCD00001581