Order Entry
Finland
ContactUsLinkComponent
4-Isopropylcyclohexanecarboxylic acid (cis and trans mixture) 95%
4-Isopropylcyclohexanecarboxylic acid (cis and trans mixture) 95%
Catalog # APOSOR931378-5G
Supplier:  Apollo Scientific
CAS Number:  
4-Isopropylcyclohexanecarboxylic acid (cis and trans mixture) 95%
Catalog # APOSOR931378-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :170.25 g/mol
Storage Temperature:Ambient
CAS Number:62067-45-2
Formula:C₁₀H₁₈O₂
MDL Number:MFCD01734696