Order Entry
Finland
ContactUsLinkComponent
N,N-Diethyl-1,4-cyclohexanediamine (cis and trans mixture) 95%
N,N-Diethyl-1,4-cyclohexanediamine (cis and trans mixture) 95%
Catalog # APOSOR929777-1G
Supplier:  Apollo Scientific
CAS Number:  
N,N-Diethyl-1,4-cyclohexanediamine (cis and trans mixture) 95%
Catalog # APOSOR929777-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :170.30 g/mol
Storage Temperature:Ambient
CAS Number:42389-54-8
Formula:C₁₀H₂₂N₂
MDL Number:MFCD06658428