Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Methyloxindole
4-Methyloxindole 98%
4-Methyloxindole 98%
4-Methyloxindole 98%
Supplier:
Apollo Scientific
Certificates
Expand image
4-Methyloxindole 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
13220-46-7
:
Formula:
C₉H₉NO
MDL Number:
MFCD09835631
:
147.177 g/mol
Storage Temperature:
Ambient