3-Acetyl-2,4-dimethylpyrrole 95%
Catalog # APOSOR926294-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:137.18 g/mol
Storage Temperature:Ambient
CAS Number:2386-25-6
Formula:C₈H₁₁NO
MDL Number:MFCD00005221