Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
cis-Jasmone
cis-Jasmone 95%
cis-Jasmone 95%
APOSOR925069-100G
:
Apollo Scientific
:
cis-Jasmone 95%
APOSOR925069-100G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
100 g
Seq:
3401
:
:
164.25 g/mol
:
Ambient
:
488-10-8
:
C₁₁H₁₆O
:
MFCD00001402