0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Finland
ContactUsLinkComponent
1-Ethyl-1H-pyrrole-2-carboxaldehyde
1-Ethyl-1H-pyrrole-2-carboxaldehyde 95%
1-Ethyl-1H-pyrrole-2-carboxaldehyde 95%
1-Ethyl-1H-pyrrole-2-carboxaldehyde 95%
Supplier:
Apollo Scientific
Certificates
Expand image
1-Ethyl-1H-pyrrole-2-carboxaldehyde 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
2167-14-8
:
Formula:
C₇H₉NO
MDL Number:
MFCD11909543
:
123.16 g/mol
Storage Temperature:
Ambient