4-Butylcyclohexanol (cis- and trans- mixture) 95%
Catalog # APOSOR920133-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:156.27 g/mol
Storage Temperature:Ambient
CAS Number:70568-60-4
Formula:C₁₀H₂₀O
MDL Number:MFCD01321147