

tert-Butyl-4-[4-(aminomethyl)phenyl]tetrahydro-1(2H)-pyrazinecarboxylate 90%
APOSOR9186-1G
:
- Pack type:Bottle
- Pk:1 g
- Seq:3047
:
:291.4 g/mol
:83…87 °C
:Ambient
:2923
:8+6.1,III
:GHS05
:GHS07
:852180-47-3
:C₁₆H₂₅N₃O₂
:MFCD03791213