Order Entry
Finland
ContactUsLinkComponent
trans-Methyl 4-(hydroxymethyl)cyclohexanecarboxylate 95%
trans-Methyl 4-(hydroxymethyl)cyclohexanecarboxylate 95%
Catalog # APOSOR917667-1G
Supplier:  Apollo Scientific
CAS Number:  
trans-Methyl 4-(hydroxymethyl)cyclohexanecarboxylate 95%
Catalog # APOSOR917667-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :172.22 g/mol
Storage Temperature:Ambient
CAS Number:110928-44-4
 :13380-85-3
Formula:C₉H₁₆O₃
MDL Number:MFCD03840884