Order Entry
Finland
ContactUsLinkComponent
4-Amylcyclohexylamine (cis- and trans- mixture) 95%
4-Amylcyclohexylamine (cis- and trans- mixture) 95%
Catalog # APOSOR916545-5G
Supplier:  Apollo Scientific
CAS Number:  
4-Amylcyclohexylamine (cis- and trans- mixture) 95%
Catalog # APOSOR916545-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :169.31 g/mol
Storage Temperature:Ambient
CAS Number:38793-01-0
Formula:C₁₁H₂₃N
MDL Number:MFCD07778407