Dimethyl terephthalate 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:120-61-6
:
Formula:C₁₀H₁₀O₄
MDL Number:MFCD00008440
Boiling Pt:288 °C
Melting Pt:140…143 °C
Density:1.36 g/cm³ (25 °C)
Flash Pt:151 °C
:194.19 g/mol
Storage Temperature:Ambient