Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Amino-5-nitrobenzonitrile
2-Amino-5-nitrobenzonitrile 97%
2-Amino-5-nitrobenzonitrile 97%
Irritant
2-Amino-5-nitrobenzonitrile 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2-Amino-5-nitrobenzonitrile 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
17420-30-3
:
Formula:
C₇H₅N₃O₂
MDL Number:
MFCD00007362
:
163.14 g/mol
Melting Pt:
200…207 °C
Flash Pt:
234 °C
:
GHS07
Storage Temperature:
Ambient