Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sodium butyrate
Sodium butyrate
Sodium butyrate
Sodium butyrate
Supplier:
Apollo Scientific
Certificates
Expand image
Sodium butyrate
Supplier:
Apollo Scientific
About this item
CAS Number:
156-54-7
:
Formula:
C₄H₇NaO₂
MDL Number:
MFCD00002816
:
110.09 g/mol
Melting Pt:
250…253 °C
EINECS:
205-857-6
:
GHS07
Storage Temperature:
Ambient