Order Entry
Finland
ContactUsLinkComponent
4-Bromothiazole 97%
Corrosive
Irritant
4-Bromothiazole 97%
Catalog # APOSOR8552-25G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Irritant
4-Bromothiazole 97%
Catalog # APOSOR8552-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :164.03 g/mol
 :GHS05
 :GHS07
Storage Temperature:Refrigerator
CAS Number:34259-99-9
Formula:C₃H₂BrNS
MDL Number:MFCD06657592