
3,5-Dichlorobenzaldehyde, Technical Grade
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:10203-08-4
:
Formula:C₇H₄Cl₂O
MDL Number:MFCD00003352
:175.01 g/mol
Boiling Pt:235…240 °C
Melting Pt:62…66 °C
Storage Temperature:Ambient
EINECS:233-499-0
UN:3261
ADR:8,II
:GHS05