Order Entry
Finland
ContactUsLinkComponent
Diethyl [(Tetrahydropyran-2-yloxy)methyl]phosphonate
Irritant
Diethyl [(Tetrahydropyran-2-yloxy)methyl]phosphonate
Catalog # APOSOR8421-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Diethyl [(Tetrahydropyran-2-yloxy)methyl]phosphonate
Catalog # APOSOR8421-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :252.25 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:71885-51-3
Formula:C₁₀H₂₁O₅P
MDL Number:MFCD09263515