Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 4-bromo-3-nitrobenzoate
Methyl-4-bromo-3-nitrobenzoate
Methyl-4-bromo-3-nitrobenzoate
:
Apollo Scientific
Methyl-4-bromo-3-nitrobenzoate
:
Apollo Scientific
:
2363-16-8
:
:
C₈H₆BrNO₄
:
MFCD00553089
:
260.04 g/mol
:
104 °C
:
GHS07
:
Ambient