0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Finland
ContactUsLinkComponent
(1,5-Dimethyl-1H-pyrrol-2-yl)methylamine
(1,5-Dimethyl-1H-pyrrol-2-yl)methylamine, Technical Grade
(1,5-Dimethyl-1H-pyrrol-2-yl)methylamine, Technical Grade
:
Apollo Scientific
(1,5-Dimethyl-1H-pyrrol-2-yl)methylamine, Technical Grade
:
Apollo Scientific
:
118799-24-9
:
:
C₇H₁₂N₂
:
MFCD02180786
:
124.19 g/mol
:
40 °C/0.1mmHg
:
Refrigerator
:
2735
:
8,III
:
GHS05