Order Entry
Finland
ContactUsLinkComponent
3-Nitro-o-toluic acid
Irritant
3-Nitro-o-toluic acid
Catalog # APOSOR7971-500G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3-Nitro-o-toluic acid
Catalog # APOSOR7971-500G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :181.15 g/mol
Melting Pt:182…186 °C
EINECS:217-826-4
 :GHS07
Storage Temperature:Ambient
CAS Number:1975-50-4
Formula:C₈H₇NO₄
MDL Number:MFCD00007160