


2,4-Dichlorobenzaldehyde 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:874-42-0
:
Formula:C₇H₄Cl₂O
MDL Number:MFCD00003305
:175.01 g/mol
Boiling Pt:233 °C
Melting Pt:69…73 °C
Flash Pt:135 °C
EINECS:212-861-1
UN:1759
ADR:8,III
:GHS05
:GHS07
:GHS09
Storage Temperature:Ambient