
4-(tert-Butoxycarbonyl)benzoic acid
Catalog # APOSOR7829-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:222.24 g/mol
Melting Pt:209…212 °C (dec.)
:GHS07
Storage Temperature:Ambient
CAS Number:20576-82-3
Formula:C₁₂H₁₄O₄
MDL Number:MFCD04037901