Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Aminosalicylic acid
3-Aminosalicylic acid 97%
3-Aminosalicylic acid 97%
Irritant
3-Aminosalicylic acid 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
3-Aminosalicylic acid 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
570-23-0
:
Formula:
C₇H₇NO₃
MDL Number:
MFCD00010299
:
153.14 g/mol
Melting Pt:
∼ 240 °C (dec.)
EINECS:
209-328-0
:
GHS07
Storage Temperature:
Ambient