Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,5-Dichloro-4-pyridone
3,5-Dichloro-4-pyridone
3,5-Dichloro-4-pyridone
:
Apollo Scientific
3,5-Dichloro-4-pyridone
:
Apollo Scientific
:
17228-70-5
:
:
C₅H₃Cl₂NO
:
MFCD00051673
:
325 °C
:
163.9894 g/mol
:
241-267-5
:
GHS07
:
Ambient