
4,7-Dibromo-3-hydroxy-2-naphthoic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1779-10-8
:
Formula:C₁₁H₆Br₂O₃
MDL Number:MFCD00046367
:345.98 g/mol
Melting Pt:251…255 °C
EINECS:217-214-7
:GHS07
Storage Temperature:Ambient