Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
m-Toluic acid
m-Toluic acid
m-Toluic acid
Irritant
m-Toluic acid
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
m-Toluic acid
Supplier:
Apollo Scientific
About this item
CAS Number:
99-04-7
:
Formula:
C₈H₈O₂
MDL Number:
MFCD00002523
:
136.15 g/mol
:
GHS07
Storage Temperature:
Ambient