Order Entry
Finland
ContactUsLinkComponent
2-Methylimidazo[1,2-a]pyridine-3-carbaldehyde 97%
Irritant
2-Methylimidazo[1,2-a]pyridine-3-carbaldehyde 97%
Catalog # APOSOR6739-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-Methylimidazo[1,2-a]pyridine-3-carbaldehyde 97%
Catalog # APOSOR6739-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :160.1700 g/mol
Melting Pt:117…118 °C
 :GHS07
Storage Temperature:Refrigerator
CAS Number:30384-93-1
Formula:C₉H₈N₂O
MDL Number:MFCD02251327