
6-[2-(Pyrrolidin-1-yl)ethyl]pyridine-3-carboxylic acid 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:387350-41-6
:
Formula:C₁₂H₁₆N₂O₂
MDL Number:MFCD01319352
:220.27 g/mol
Melting Pt:198…205 °C (dec.)
:GHS07
Storage Temperature:Ambient