Order Entry
Finland
ContactUsLinkComponent
4-Chloro-1H-pyrazolo[3,4-d]pyrimidine 97%
Irritant
4-Chloro-1H-pyrazolo[3,4-d]pyrimidine 97%
Catalog # APOSOR6423-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-Chloro-1H-pyrazolo[3,4-d]pyrimidine 97%
Catalog # APOSOR6423-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :154.5571 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:5399-92-8
Formula:C₅H₃ClN₄
MDL Number:MFCD03030404