Order Entry
Finland
ContactUsLinkComponent
Methyl-3-bromo-2-oxo-2H-pyran-5-carboxylate 95%
Methyl-3-bromo-2-oxo-2H-pyran-5-carboxylate 95%
Catalog # APOSOR6402-5G
Supplier:  Apollo Scientific
CAS Number:  
Methyl-3-bromo-2-oxo-2H-pyran-5-carboxylate 95%
Catalog # APOSOR6402-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :233.02 g/mol
Storage Temperature:Ambient
CAS Number:42933-07-3
Formula:C₇H₅BrO₄
MDL Number:MFCD00463742