Order Entry
Finland
ContactUsLinkComponent
Methyl-5-bromo-2-hydroxynicotinate 97%
Irritant
Methyl-5-bromo-2-hydroxynicotinate 97%
Catalog # APOSOR6397-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Methyl-5-bromo-2-hydroxynicotinate 97%
Catalog # APOSOR6397-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :232.03 g/mol
Melting Pt:180…182 °C
 :GHS07
Storage Temperature:Refrigerator
CAS Number:120034-05-1
Formula:C₇H₆BrNO₃
MDL Number:MFCD03844843