Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
L-Propargylglycine
L-Propargylglycine
L-Propargylglycine
L-Propargylglycine
Supplier:
Apollo Scientific
Certificates
Expand image
L-Propargylglycine
Supplier:
Apollo Scientific
About this item
CAS Number:
23235-01-0
:
Formula:
C₅H₇NO₂
MDL Number:
MFCD00077855
:
113.116 g/mol
:
GHS07
Storage Temperature:
Refrigerator