

p-Cresol 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:106-44-5
:
Formula:C₇H₈O
MDL Number:MFCD00002376
:108.14 g/mol
Boiling Pt:202 °C
Melting Pt:32…34 °C
Density:1.034 g/cm³ (25 °C)
Flash Pt:85 °C
Storage Temperature:Ambient
EINECS:203-398-6
UN:3455
ADR:6.1+8,II
:GHS05
:GHS06