Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 4-methoxy-2-nitrobenzoate
Methyl-4-methoxy-2-nitrobenzoate
Methyl-4-methoxy-2-nitrobenzoate
:
Apollo Scientific
Methyl-4-methoxy-2-nitrobenzoate
:
Apollo Scientific
:
181871-73-8
:
:
C₉H₉NO₅
:
MFCD06658482
:
211.17 g/mol
:
GHS07
:
Ambient