Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 4-amino-3-methylbenzoate
Methyl-4-amino-3-methylbenzoate 98%
Methyl-4-amino-3-methylbenzoate 98%
Irritant
Methyl-4-amino-3-methylbenzoate 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
Methyl-4-amino-3-methylbenzoate 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
18595-14-7
:
Formula:
C₉H₁₁NO₂
MDL Number:
MFCD00102230
:
165.19 g/mol
Melting Pt:
116…120 °C
:
GHS07
Storage Temperature:
Ambient