1,5-Dimethyl-1H-indole-2-carbaldehyde 95+%
Catalog # APOSOR5667-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:173.22 g/mol
Storage Temperature:Ambient
CAS Number:883526-76-9
Formula:C₁₁H₁₁NO
MDL Number:MFCD06800611