0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Finland
ContactUsLinkComponent
2-Methylpropane
2-Methylpropane
2-Methylpropane
Flammable
2-Methylpropane
Supplier:
Apollo Scientific
Certificates
Expand image
Flammable
2-Methylpropane
Supplier:
Apollo Scientific
About this item
CAS Number:
75-28-5
:
Formula:
C₄H₁₀
:
58.12 g/mol
Boiling Pt:
–12 °C
Melting Pt:
–160 °C
Flash Pt:
–83 °C
Storage Temperature:
Ambient
UN:
1969
ADR:
2.1,
:
GHS02