Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-(Boc-amino)cyclohexanone
4-(Boc-amino)cyclohexanone 98%
4-(Boc-amino)cyclohexanone 98%
Irritant
4-(Boc-amino)cyclohexanone 98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4-(Boc-amino)cyclohexanone 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
179321-49-4
:
Formula:
C₁₁H₁₉NO₃
MDL Number:
MFCD00798168
:
213.2700 g/mol
:
GHS07
Storage Temperature:
Ambient