Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Bromo-4'-nitrobiphenyl
4-Bromo-4'-nitrobiphenyl 95+%
4-Bromo-4'-nitrobiphenyl 95+%
APOSOR53160-1G
:
Apollo Scientific
:
4-Bromo-4'-nitrobiphenyl 95+%
APOSOR53160-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
278.1014 g/mol
:
GHS07
:
Ambient
:
6242-98-4
:
C₁₂H₈BrNO₂
:
MFCD00024697