Order Entry
Finland
ContactUsLinkComponent
N,N-Dimethyl-1,4-phenylenediammonium dichloride 95%
N,N-Dimethyl-1,4-phenylenediammonium dichloride 95%
Catalog # APOSOR5247-100G
Supplier:  Apollo Scientific
CAS Number:  
N,N-Dimethyl-1,4-phenylenediammonium dichloride 95%
Catalog # APOSOR5247-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :209.12 g/mol
Melting Pt:222 °C (dec.)
Storage Temperature:Refrigerator
EINECS:208-635-7
UN:2811
ADR:6.1,II
 :GHS06
 :GHS07
CAS Number:536-46-9
 :MFCD00012990
Formula:(CH₃)₂NC₆H₄NH₂·2HCl
MDL Number:MFCD00012991