

(S)-(-)-2-Aminomethyl-1-ethylpyrrolidine 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:22795-99-9
:
Formula:C₇H₁₆N₂
MDL Number:MFCD00191371
Boiling Pt:50…52 °C
Melting Pt:57 °C
UN:2733
ADR:3+8,III
:128.22 g/mol
:GHS02
:GHS05
Storage Temperature:Ambient