Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,4,5-Trimethoxybenzylamine
3,4,5-Trimethoxybenzylamine 96%
3,4,5-Trimethoxybenzylamine 96%
:
Apollo Scientific
3,4,5-Trimethoxybenzylamine 96%
:
Apollo Scientific
:
18638-99-8
:
:
C₁₀H₁₅NO₃
:
MFCD00017150
:
197.23
:
120…122 °C/0.05mmHg
:
1.155 g/cm³ (25 °C)
:
>110 °C
:
Ambient
:
242-468-0
:
2735
:
8,III
:
GHS05