Order Entry
Finland
Orders LinkContactUsLinkComponent
(S)-(-)-1,2-Epoxypropane 97%

(S)-(-)-1,2-Epoxypropane 97%

Supplier:   Apollo Scientific
(S)-(-)-1,2-Epoxypropane 97%
Supplier:  Apollo Scientific
Certificates
(S)-(-)-1,2-Epoxypropane 97%

2 Options Available Below

Product Details & Documents
Synonyms:  (S)-(-)-Methyloxirane, (S)-(-)-Propylene oxide, (S)-(-)-Propene oxide
  • CAS Number:16088-62-3
  • Formula:C₃H₆O
  • MDL Number:MFCD00064312
  • Molecular Weight:58.08 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g

  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g