Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(2,5-Dichloro-4-pyridinyl)methanol
(2,5-Dichloro-4-pyridinyl)methanol 97%
(2,5-Dichloro-4-pyridinyl)methanol 97%
(2,5-Dichloro-4-pyridinyl)methanol 97%
Supplier:
Apollo Scientific
Certificates
Expand image
(2,5-Dichloro-4-pyridinyl)methanol 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
866039-42-1
:
Formula:
C₆H₅Cl₂NO
MDL Number:
MFCD04125666
:
178.02 g/mol
Storage Temperature:
Ambient