Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Fmoc-4-cyano-D-phenylalanine
Fmoc-4-cyano-D-phenylalanine 95%
Fmoc-4-cyano-D-phenylalanine 95%
Fmoc-4-cyano-D-phenylalanine 95%
APOSOR471678-5G
:
Apollo Scientific
:
Fmoc-4-cyano-D-phenylalanine 95%
APOSOR471678-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
412.45 g/mol
:
Ambient
:
205526-34-7
:
C₂₅H₂₀N₂O₄
:
MFCD00672560