4-methoxy-1-naphthoic acid 98%
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: 4-Methoxynaphthalene-1-carboxylic acid, 4-Methoxy-1-naphthoic acid
- CAS Number:13041-62-8
- Formula:C₁₂H₁₀O₃
- MDL Number:MFCD02575372
- Molecular Weight:202.209 g/mol
- Storage Temperature:Ambient
3 Options Available Below
Product Details & Documents
Synonyms: 4-Methoxynaphthalene-1-carboxylic acid, 4-Methoxy-1-naphthoic acid
- CAS Number:13041-62-8
- Formula:C₁₂H₁₀O₃
- MDL Number:MFCD02575372
- Molecular Weight:202.209 g/mol
- Storage Temperature:Ambient
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



