Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Amino-2-cyclohexen-1-one
3-Amino-2-cyclohexen-1-one 95%
3-Amino-2-cyclohexen-1-one 95%
Irritant
3-Amino-2-cyclohexen-1-one 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
3-Amino-2-cyclohexen-1-one 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
5220-49-5
:
Formula:
C₆H₉NO
MDL Number:
MFCD00013783
:
111.14 g/mol
Melting Pt:
129…135 °C
EINECS:
226-014-9
:
GHS07
Storage Temperature:
Ambient