
1-Acetyl-4-piperidone 95+%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:32161-06-1
:
Formula:C₇H₁₁NO₂
MDL Number:MFCD00006190
:141.17 g/mol
Boiling Pt:152 °C/0.75mmHg
Density:1.15 g/cm³ (25 °C)
Flash Pt:112 °C
:GHS07
Storage Temperature:Ambient